Learn More
Revvity Health Sciences Inc Triiiodothyronine, L-3, 5, 3'-[125I], 250 µCi (9.25 MBq), Specific Activity: 3300 µCi (122 MBq)/µg
Supplier:  Revvity Health Sciences Inc NEX110X250UC
Encompass
Triiiodothyronine, L-3, 5, 3'-[125I], 250 µCi (9.25 MBq), Specific Activity: 3300 µCi (122 MBq)/µg
Catalog No. 50-281-1010