Learn More
Medchemexpress LLC Triiodothyronine sulfate | 31135-55-4 | 98.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY126996100MG
Encompass_Preferred
Triiodothyronine sulfate is the main metabolite of the thyroid hormone triiodothyronine (T3). T3 is an active form of thyroid hormone that binds to the β1 thyroid hormone receptor (TRβ1) and activates its activity. In vitro, Triiodothyronine (T3, 100 nM) stimulates the proliferation of hepatocarcinoma cells when TRβ1 is overexpressed. It promotes growth, induces differentiation, and regulates metabolic effects.
  • Main metabolite of thyroid hormone triiodothyronine (T3)
  • Binds to β1 thyroid hormone receptor (TRβ1)
  • Activates TRβ1 activity
  • Stimulates proliferation of hepatocarcinoma cells (in vitro)
  • Promotes growth and induces differentiation
  • Regulates metabolic effects
Catalog No. 50-003-42825