Learn More
Medchemexpress LLC Triiodothyronine sulfate | 31135-55-4 | 98.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12699650MG
Encompass_Preferred
Triiodothyronine sulfate is the main metabolite of thyroid hormone triiodothyronine (T3). Triiodothyronine is an active form of thyroid hormone, which binds to β1 thyroid hormone receptor (TRβ1), and activates its activity.
  • Stimulates the proliferation of hepatocarcinoma cells where TRβ1 is overexpressed.
  • Binds to human β1 thyroid hormone receptor (hTRβ1) and changes its conformation.
  • Promotes growth, induces differentiation, and regulates metabolic effects.
Catalog No. 50-003-40824