Learn More
Medchemexpress LLC HY-101467 5mg , Trilaciclib CAS:1374743-00-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1014675MG
Encompass_Preferred
HY-101467 5mg Trilaciclib CAS: 1374743-00-6 Trilaciclib is a CDK4/6 inhibitor with IC50s of 1 nM and 4 nM for CDK4 and CDK6, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1250