Learn More
Medchemexpress LLC Trilobatin | 4192-90-9 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN4100
Encompass_Preferred
Trilobatin is a natural sweetener derived from Lithocarpus polystachyus Rehd. It acts as an HIV-1 entry inhibitor by targeting the HIV-1 Gp41 envelope and exhibits neuroprotective effects. Additionally, Trilobatin is an SGLT1/2 inhibitor that selectively induces the proliferation of human hepatoblastoma cells.
  • Derived from Lithocarpus polystachyus Rehd
  • HIV-1 entry inhibitor
  • Exhibits neuroprotective effects
  • Functions as an SGLT1/2 inhibitor
  • Induces proliferation of human hepatoblastoma cells
  • Protects against oxidative injury in neuronal PC12 cells
Catalog No. 50-004-21857