Learn More
Medchemexpress LLC Trilobatin | 4192-90-9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYN410050MG
Encompass_Preferred
Trilobatin is a natural sweetener derived from Lithocarpus polystachyus Rehd. It functions as an HIV-1 entry inhibitor by targeting the HIV-1 Gp41 envelope and exhibits neuroprotective effects. Additionally, Trilobatin is an SGLT1/2 inhibitor that selectively induces the proliferation of human hepatoblastoma cells.
  • Functions as an HIV-1 entry inhibitor
  • Targets the HIV-1 Gp41 envelope
  • Exhibits neuroprotective effects
  • Acts as an SGLT1/2 inhibitor
  • Induces the proliferation of human hepatoblastoma cells
  • Protects against oxidative injury in neuronal PC12 cells
  • Regulates mitochondrial reactive oxygen species (mtROS) homeostasis
Catalog No. 50-004-20634