Learn More
Medchemexpress LLC Trilobatin | 4192-90-9 | MFCD16621247 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN41001MG
Encompass_Preferred
Trilobatin is a natural sweetener derived from Lithocarpus polystachyus Rehd. It acts as an HIV-1 entry inhibitor by targeting the HIV-1 Gp41 envelope. This compound also exhibits neuroprotective effects and is an SGLT1/2 inhibitor that selectively induces the proliferation of human hepatoblastoma cells. It is intended for research use only.
  • Derived from plants Euphorbiaceae Vernicia fordii and Fagaceae
  • Acts as an HIV-1 entry inhibitor
  • Targets HIV-1 Gp41 envelope
  • Exhibits neuroprotective effects
  • Functions as an SGLT1/2 inhibitor
  • Selectively induces proliferation of human hepatoblastoma cells
  • Protects against oxidative injury in neuronal PC12 cells
  • Regulates mitochondrial reactive oxygen species (mtROS) homeostasis
Catalog No. 50-004-18543