Learn More
MSC Vermont Gage, Class ZZ Plus Plug Gage, 1.3114 in. Diameter, 1 ea.
Supplier:  MSC 64760259
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications, featuring a diameter of 1.3114 in. It is engineered for reliable inspection and quality control tasks within metalworking and industrial environments.
  • Diameter of 1.3114 in.
  • Class ZZ Plus tolerance
  • Handle sold separately
Catalog No. 50-005-47875