Learn More
Medchemexpress LLC Trilostane (Win 24540) | 13647-35-3 | 99.9% | C20H27NO3 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY14281200MG
Encompass_Preferred
Trilostane (Win 24540) is a competitive and orally active 3-β-hydroxysteroiddehydrogenase (3β-HSD) inhibitor. It is a synthetic nonhormonal steroid that can be used for the research of breast cancer and prostate cancer.
  • Competitive and orally active 3-β-hydroxysteroiddehydrogenase (3β-HSD) inhibitor.
  • Synthetic nonhormonal steroid.
  • Used for the research of breast cancer and prostate cancer.
  • Influences pregnenolone metabolism in adrenal cortex.
  • Selectively inhibits pregnenolone converting to progesterone in the adrenal gland.
  • Effectively controls pituitary-dependent hyperadrenocorticism (PDH) in dogs.
Catalog No. 50-003-53618