Learn More
Aviva Systems Biology Corp TRIM15 Antibody - N-terminal region (ARP34645_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34645_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM15. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TRIM15. Alternate names: RNF93, ZNFB7, ZNF178ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55498