Learn More
Aviva Systems Biology Corp TRIM68 Antibody - C-terminal region (ARP34460_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34460_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TRIM68. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TRIM68. Alternate names: SS56, GC109, SS-56, RNF137ApplicationIHC, WB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-55401