Learn More
Genetex Inc TRIM72 Monoclonal Antibody [HMV313] HistoMAX™ (GTX639937) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639937
Encompass_Preferred
The TRIM72 antibody [HMV313] HistoMAX™ (GTX639937) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human rhabdomyosarcoma (RMS) tissue using GTX639937 TRIM72 antibody [HMV313] HistoMAX. Rhabdomyosarcoma with strong TRIM72 immunostaining of tumor cells.
Catalog No. NC3669313