Learn More
Selleck Chemical LLC Trimetazidine dihydrochloride S4543-1g
SDP
Supplier:  Selleck Chemical LLC S45431G
Encompass_Preferred
Trimetazidine dihydrochloride (Yoshimilon Kyurinett Vastarel F) is a kind of drug for treatment of chronic ischemic disorders It improves myocardial glucose utilization through inhibition of fatty acid metabolism known as fatty acid oxidation inhibitor
Catalog No. 50-311-1024