Learn More
Medchemexpress LLC Trimethobenzamide hydrochloride | 554-92-7 | 99.9% | 200 MG
SDP
Supplier:  Medchemexpress LLC HY12751A200MG
Encompass_Preferred
Trimethobenzamide hydrochloride is a non-phenothiazine benzamide antiemetic that acts centrally to block D2 receptors. It inhibits the medullary chemoreceptor trigger zone, blocking emetic impulses to prevent nausea and vomiting.
  • Acts as a blocker of the D2 receptor
  • Used to prevent nausea and vomiting
  • Exhibits 60% to 100% oral bioavailability
  • Achieves peak effect approximately 45 minutes after oral administration
  • Achieves peak effect approximately 30 minutes after intramuscular administration
Catalog No. 50-003-40044