Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimethoprim | 738-70-5 | MFCD00036761 | 100.0% | 250 MG
SDP

Supplier:  Medchemexpress LLC 250MGHYB0510R

Encompass_Preferred

Trimethoprim is a bacteriostatic antibiotic that functions as an orally active dihydrofolate reductase inhibitor. It demonstrates effectiveness against a broad spectrum of both Gram-positive and Gram-negative aerobic bacteria, with potential applications in treating various infections.

  • Inhibits dihydrofolate reductase (DHFR) activity.
  • Effective against Gram-positive and Gram-negative aerobic bacteria.
  • Researched for urinary tract infections, Shigellosis, and Pneumocystis pneumonia.
  • Inhibits Influenza A virus infection in chick embryos when combined with zinc.
  • Molecular weight: 290.32.
  • Formula: C14H18N4O3.
  • Appearance: Solid, white to off-white.
  • Soluble in DMSO (50 mg/mL) and H2O (0.67 mg/mL).
  • Induces protein aggregation and heat shock proteins in E. coli cells.
  • Shows antibacterial activity in vivo against various bacteria.

Catalog No. 50-004-07434


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.