Learn More
TOKU-E Trimethoprim ReadyMade™ Solution | 25 mg/ML in DMSO | filter-sterile | 1 ML
Supplier:  TOKU-E T1441ML
Encompass_Preferred
Trimethoprim ReadyMade™ Solution | 25 mg/ML in DMSO | filter-sterile | 1 ML
Catalog No. NC2068795