Learn More
Strem, An Ascensus Company CAS# 1271-07-4. 500mg. (Trimethyl)cyclopentadienylplatinum(IV), 99%. MFCD07369041
SDP
Supplier:  Strem, An Ascensus Company 781300500MG
Encompass
CAS# 1271-07-4. 500mg. (Trimethyl)cyclopentadienylplatinum(IV), 99%. MFCD07369041. Molecular Weight: 305.28. Molecular Formula: (CH3)3(C5H5)Pt. Color/form: white to off-white pwdr. Strem# 78-1300. http://www.strem.com/catalog/v/78-1300/
Catalog No. 50-901-13236