Learn More
Medchemexpress LLC Trimethylacetohydrazideammonium chloride | 123-46-6 | MFCD00012009 | 99.8% | 167.64 g/mol | C5H14ClN3O | 500 G
SDP
Supplier:  Medchemexpress LLC HYW074541500G
Encompass_Preferred
Girard's reagent T (trimethylacetohydrazideammonium chloride) is a crystalline derivatizing agent used to convert aldehydes and ketones into stable hydrazones for chromatographic and spectrophotometric analysis. It is applied in analytical workflows and in synthetic chemistry for preparing pharmaceutical and agrochemical intermediates.
  • Derivatizes aldehydes and ketones to form stable hydrazones for improved detection.
  • Used in analytical and synthetic chemistry applications.
  • Solid crystalline form, soluble in alcohols and ethers.
  • CAS number 123-46-6; chemical formula C5H14ClN3O; molecular weight 167.64 g/mol.
  • High purity, reported as 99.83% on the product listing.
Catalog No. 50-002-10815