Learn More
Cayman Chemical TrImethylamIn-d9 N-oxIde 10mg
SDP
Supplier:  Cayman Chemical 3144910 MG
Encompass_Preferred
An internal standard for the quantification of trimethylamine N-oxide by GC- or LC-MS
Catalog No. 50-297-8909