Learn More
Medchemexpress LLC Trimethylamine N-oxi 500mg | 1184-78-7 | 75.11 g/mol | C3H9NO | 500 MG
SDP
Supplier:  Medchemexpress LLC HY116084500MG
Encompass_Preferred
Trimethylamine N-oxide is a small tertiary amine oxide used as a research reagent and analytical standard in biochemical and metabolic studies. It is provided in both solid and solution formats for laboratory research and analytical applications.
  • Available as solid and as 10 mM solution in DMSO.
  • High purity (>98.0%).
  • Molecular weight 75.11 g/mol; formula C3H9NO.
  • White to off-white solid with recommended cold storage for stability.
  • Intended for research and analytical applications involving gut-microbiota metabolites.
Catalog No. 50-002-29984