Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimethylamine N-oxi 5g | 1184-78-7 | 5 G
SDP

Supplier:  Medchemexpress LLC HY1160845G

Encompass_Preferred

Trimethylamine N-oxide is a research-grade small-molecule reagent (CAS 1184-78-7) provided as a solid with a reported purity of 98.0%. It is used in studies of gut microbiome-derived metabolites and inflammatory pathways such as ROS and NLRP3 inflammasome activation.

  • Solid form suitable for laboratory use
  • Reported purity 98.0%
  • Used to study gut microbiome metabolites and inflammatory signaling
  • Available in multiple pack sizes including 5 g
  • CAS number 1184-78-7 for unambiguous identification

Catalog No. 50-002-27272


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.