Learn More
Medchemexpress LLC Trimethylamine N-oxide dihydrate | 62637-93-8 | >98.0% | 111.14 | 10 MG
SDP
Catalog No. 5000344617
Encompass_Preferred
Trimethylamine N-oxide dihydrate is a gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients. It induces inflammation by activating the ROS/NLRP3 inflammasome. Additionally, it accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF-β/smad2 signaling pathway.
  • Gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients.
  • Induces inflammation by activating the ROS/NLRP3 inflammasome.
  • Accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF-β/smad2 signaling pathway.
  • Can be used in animal modeling to create cardiovascular and metabolic disease models.
  • Found in tissues of various marine organisms, protecting against adverse effects of temperature, salinity, high urea, and hydrostatic pressure.
Catalog No. 50-003-44617 Supplier Medchemexpress LLC Supplier No. HY108915100G
Trimethylamine N-oxide dihydrate is a gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients. It induces inflammation by activating the ROS/NLRP3 inflammasome. Additionally, it accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF-β/smad2 signaling pathway.
  • Gut microbe-dependent metabolite of dietary choline and other trimethylamine-containing nutrients.
  • Induces inflammation by activating the ROS/NLRP3 inflammasome.
  • Accelerates fibroblast-myofibroblast differentiation and induces cardiac fibrosis by activating the TGF-β/smad2 signaling pathway.
  • Can be used in animal modeling to create cardiovascular and metabolic disease models.
  • Found in tissues of various marine organisms, protecting against adverse effects of temperature, salinity, high urea, and hydrostatic pressure.