Learn More
Medchemexpress LLC Trimethyllysine-d9 | 1182037-78-0 | 95.0% | C9H11D9N2O2 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HY143711S2.5MG
Encompass_Preferred
Trimethyllysine-d9 is the deuterium-labeled Trimethyllysine. Trimethyllysine is a post-translationally modified amino acid involved in carnitine biosynthesis and epigenetic processes. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-54412