Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimethylolpropane ethoxylate triacrylate | 28961-43-5 | 428.47 | 100 G
SDP

Supplier:  Medchemexpress LLC HY139446100G

Encompass_Preferred

Trimethylolpropane ethoxylate triacrylate (ETPTA) is a plasticizer that can be used to synthesize nanocomposites. It is intended for research use only.

  • Used as a plasticizer for synthesizing nanocomposites.
  • Suitable for laboratory chemical applications.
  • Appears as a colorless to light yellow liquid.
  • Soluble in DMSO (≥ 100 mg/mL).
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
  • Ships at room temperature if less than 2 weeks.
  • Can be used in fabricating photonic crystals.

Catalog No. 50-003-49486


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.