Learn More
Medchemexpress LLC Trimethylphenylammonium bromide | 16056-11-4 | 98.0% | 216.12 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW012450
Encompass_Preferred
Trimethylphenylammonium bromide is a quaternary ammonium salt consisting of a positively charged N,N,N-trimethylanilinium cation and a negatively charged bromide anion. It is commonly used as a phase transfer catalyst in organic chemical reactions, facilitating the transfer of reactants between immiscible phases. It can also be used as a reagent in the synthesis of various organic compounds and in some applications as a surfactant or corrosion inhibitor. It is a biochemical reagent for life science related research.
  • Quaternary ammonium salt
  • Used as a phase transfer catalyst in organic chemical reactions
  • Facilitates transfer of reactants between immiscible phases
  • Can be used as a reagent in the synthesis of various organic compounds
  • Applicable as a surfactant or corrosion inhibitor
  • Biochemical reagent for life science related research
Catalog No. 50-004-52696