Learn More
Medchemexpress LLC Trimethylphenylammonium bromide | 16056-11-4 | 98.0% | 216.12 | 500 G
Supplier: Medchemexpress LLC HYW012450500G
Trimethylphenylammonium bromide is a quaternary ammonium salt composed of a positively charged N,N,N-trimethylanilinium cation and a negatively charged bromide anion. It is frequently employed as a phase transfer catalyst in organic chemical reactions, aiding in the transfer of reactants between immiscible phases.
- Serves as a reagent in the synthesis of various organic compounds.
- Can be utilized as a surfactant or corrosion inhibitor in certain applications.
- For research use only.