Learn More
Strem, An Ascensus Company Trimethyltin hydroxide,98% 10g
SDP
Supplier:  Strem, An Ascensus Company 93508510G
Encompass
CAS# 56-24-6. 10g. Trimethyltin hydroxide, 98%. MFCD00013929. Molecular Weight: 180.80. Molecular Formula: (CH3)3SnOH. Color/form: white pwdr. Strem# 93-5085. http://www.strem.com/catalog/v/93-5085/
Catalog No. 50-901-15637