Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Trimetrexate glucuronate | 82952-64-5 | 98.1% | 563.56 | C25H33N5O10 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY10339650MG

Encompass_Preferred

Trimetrexate glucuronate is the mono-D-glucuronate conjugate of trimetrexate, an antifolate compound used in biochemical and pharmacological research. It is supplied as a research reagent and analytical standard for studies of antifolate activity and related metabolic profiling.

  • High reported purity of 98.09% for reliable experimental use.
  • Molecular weight 563.56 g/mol for stoichiometric calculations.
  • Chemical formula C25H33N5O10 for structure-based applications.
  • Supplied as a 50 mg sample suitable for laboratory assays.
  • Commonly used as an analytical standard and research reagent in antifolate and antimicrobial studies.

Catalog No. 50-002-45048


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.