Learn More
Medchemexpress LLC Trimetrexate glucuronate | 82952-64-5 | MFCD01725300 | 98.1% | 563.56 | C25H33N5O10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10339610MG
Encompass_Preferred
Trimetrexate glucuronate is a folic acid antagonist that inhibits dihydrofolate reductase, disrupting DNA and RNA synthesis. It is used in research on antifolate mechanisms, antitumour activity, and Pneumocystis jirovecii (carinii) pneumonia.
  • Antifolate dihydrofolate reductase inhibitor.
  • Potential antitumour activity and use in Pneumocystis research.
  • Molecular formula C25H33N5O10 and molecular weight 563.56.
  • Typical supplier-specified purity ~98.1%.
  • Available in small research quantities (for example, 5 mg, 10 mg, 50 mg).
Catalog No. 50-002-44799