Learn More
Medchemexpress LLC Trimetrexate glucuronate | 82952-64-5 | MFCD01725300 | 98.1% | 563.56 g·mol⁻¹ | C25H33N5O10 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1033965MG
Encompass_Preferred
Trimetrexate glucuronate is a folic acid antagonist that inhibits dihydrofolate reductase, interfering with DNA and RNA synthesis by preventing purine and thymidylate formation. It is supplied as a research reagent with reported purity of 98.09% and is provided as an off-white to light yellow solid. Recommended storage is 4°C in a sealed container; soluble in DMSO at 100 mg/mL (needs ultrasonic).
  • inhibits dihydrofolate reductase (DHFR) and blocks nucleotide synthesis.
  • reported purity of 98.09% for analytical and research use.
  • solid form, off-white to light yellow appearance.
  • soluble in DMSO (100 mg/mL) with ultrasonic assistance.
  • recommended sealed storage at 4°C, protected from moisture.
Catalog No. 50-002-44715