Learn More
Cayman Chemical Trimidox hydrochloride
SDP
Supplier:  Cayman Chemical 100111245 MG
Encompass_Preferred
A specific ribonucleotide reductase inhibitor; inhibits growth of human promyelocytic leukemia HL-60 cells (IC50 = 35 µM)
Catalog No. 50-288-9220