Learn More
Cayman Chemical TrImIpramIn-d3maleate 5mg
SDP
Supplier:  Cayman Chemical 319195 MG
Encompass_Preferred
An internal standard for the quantification of trimipramine by GC- or LC-MS
Catalog No. 50-294-6553