Learn More
MSC ECHO Trimmer Head, Tap-to-Advance, .095 in. Line, Durable 2-Piece Construction
Supplier:  MSC 72354988
Encompass
The ECHOmatic Pro Trimmer Head is designed for professional landscapers and homeowners. It features a convenient tap-to-advance line feed mechanism and durable two-piece construction, making it ideal for medium to heavy-duty trimming applications. This head accepts up to .130 inch diameter line and comes pre-loaded with .095 inch diameter Cross-Fire line.
  • Tap-to-advance line feed for continuous operation.
  • Pre-loaded with .095 inch diameter Cross-Fire line.
  • Durable 2-piece construction for extended lifespan.
  • Designed for medium to heavy-duty applications.
  • Fits all SRM models.
  • Maximum capacity for .130 inch diameter line.
  • Product width is 6 inches.
  • Easy line loading.
Catalog No. 50-283-8619