Learn More
DIAGNOSTICA STAGO INC TriniCLOT Automated aPTT, Lyophilized Reagent For Coagulation, 10 x 6 mL
Supplier:  DIAGNOSTICA STAGO INC T1205
Encompass
The TriniCLOT Automated aPTT is a lyophilized reagent designed for the Activated Partial Thromboplastin Time assay, aiding in intravenous heparin monitoring and intrinsic factor deficiencies. Its unique composition of Platelet Factor 3 and silica activator ensures optimal sensitivity while being compatible with standard coagulation analyzers, making it a reliable choice for laboratories.
Catalog No. NC2027859
 Restricted Item