Learn More
Electron Microscopy Sciences EMS Metallurgical Trinocular Inverted Microscope
SDP
Supplier:  Electron Microscopy Sciences MET233
Encompass_Preferred
EMS Metallurgical Microscopes use long working distance objectives and reflected or incident illumination to deliver sharp, high contrast images of solid specimens. The large collector lens and light path combine with the specifically positioned diaphragms to allow more light for brighter images. A polarizer and analyzer and color filters are built-in to enhance observation. The Inverted model utilizes incident illumination for over size samples. The trinocular head allows for quick and easy attachment of cameras for documentation.
Catalog No. 50-343-49