Learn More
Medchemexpress LLC Trioctyl trimellitate | 3319-31-1 | 546.78 | 100 G
SDP
Supplier:  Medchemexpress LLC HYW010750100G
Encompass_Preferred
Trioctyl trimellitate (TOTM) is a plasticizer. It increases material plasticity by interacting with PVC molecular chains and is promising for research on the safety of medical device materials.
  • Plasticizer
  • Increases material plasticity by interacting with PVC molecular chains
  • Promising for research of the safety of medical device materials
Catalog No. 50-003-66322