Learn More
Medchemexpress LLC Tripentadecanoin-d29 | 352431-43-7 | 99.2% | 852.78 | 1 MG

Supplier: Medchemexpress LLC HY146726S1MG
Tripentadecanoin-d29 is the deuterium labeled Tripentadecanoin. It can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
- Purity of 99.2% (HPLC)
- Isotopic enrichment of 98.83%
- Stable under recommended storage conditions
- Store at -20°C, protected from light, and under nitrogen
- If in solvent, store at -80°C for 6 months or -20°C for 1 month
- Soluble in ethanol at 2 mg/mL with warming and sonication
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.