Learn More
Medchemexpress LLC Triphenyl(propyl)phosphonium-d5 (bromide) | 1703757-22-5 | 99% | 390.31 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW008678S10MG
Encompass_Preferred
Triphenyl(propyl)phosphonium-d5 (bromide) is the deuterium labeled Triphenyl(propyl)phosphonium bromide. It is intended for research use only and not for sale to patients. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules, primarily as tracers for quantitation during drug development. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52157