Learn More
Medchemexpress LLC Triphenyl(propyl)phosphonium-d5 bromide | 1703757-22-5 | 99% | 390.31 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW008678S5MG
Encompass_Preferred
Triphenyl(propyl)phosphonium-d5 (bromide) is the deuterium labeled Triphenyl(propyl)phosphonium bromide. Deuterium labeling has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs. Stable heavy isotopes like hydrogen and carbon are incorporated into drug molecules as tracers for quantitation during drug development.
  • This compound can be used as a tracer.
  • It can serve as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51277