Learn More
Medchemexpress LLC Triphenylene-d12 | 17777-56-9 | 98% | 240.36 | 50 MG
Supplier: Medchemexpress LLC 50MGHYW017708S
Triphenylene-d12 is the deuterium labeled triphenylene, intended for research purposes only. This solid compound, appearing white to off-white, can be utilized as a tracer or an internal standard for quantitative analysis. Stable heavy isotopes, such as deuterium, are incorporated into drug molecules for quantitation during drug development and may influence pharmacokinetic and metabolic profiles.
- Can be used as a tracer
- Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS