Learn More
Medchemexpress LLC Triphenylethylene | 58-72-0 | MFCD00004765 | 99.9% | 256.34 g/mol | C20H16 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1507525G
Encompass_Preferred
Triphenylethylene is an aromatic hydrocarbon with weak estrogenic and antiestrogenic activity, provided as a high-purity research reagent for laboratory studies. It is a white to light-yellow crystalline solid commonly used as a reference compound in estrogen receptor and structure-activity investigations. Follow the safety data sheet for storage and handling instructions.
  • High purity suitable for research applications.
  • White to light-yellow crystalline solid for solid-state studies.
  • Useful as an estrogen receptor and antiestrogen reference compound.
  • Supplied in a small laboratory pack size for convenience.
  • Includes COA and SDS documentation for traceability and safe handling.
Catalog No. 50-002-82375