Learn More
Cayman Chemical TrIphenylphosphIn oxIde 50mg
SDP
Supplier:  Cayman Chemical 900028950 MG
Encompass_Preferred
A coordinating solvent used to activate crystallization of chemical compounds; used in flame retardant applications, as an epoxy cure catalyst, and to produce nanostructures
Catalog No. 50-289-6294