Learn More
Medchemexpress LLC Triphenylphosphonium-derivatized anethole dithiolethione | 1429061-80-2 | 96.4% | 721.77 g/mol | C37H38BrO2PS3 | 10MG
SDP
Supplier:  Medchemexpress LLC HY12612410MG
Encompass_Preferred
AP39 is a mitochondria-targeted hydrogen sulfide (H2S) donor, a triphenylphosphonium-derivatized anethole dithiolethione used in preclinical research to raise mitochondrial H2S levels and protect against oxidative damage. It has demonstrated cytoprotective activity and preservation of mitochondrial DNA in cellular and animal models of ischemia and neurodegeneration.
  • Mitochondria-targeted delivery via a lipophilic cationic moiety.
  • Releases hydrogen sulfide (H2S) within mitochondria.
  • Exhibits cytoprotective effects under oxidative stress.
  • Preserves mitochondrial DNA integrity in preclinical studies.
  • Recommended storage and handling per supplier documentation.
Catalog No. 50-002-35015