Learn More
Sigma Aldrich Fine Chemicals Biosciences Triphosgene reagent grade500G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 330752100G
Encompass
Triphosgene [Bis(trichloromethyl)carbonate] is a versatile organic reagent used in organic synthesis alternative to phosgene. A catalytic amount of triphosgene is particularly used in chloroformylations, carbonylations, chlorinations, and dehydration reactions.
Catalog No. 50-290-6866