Learn More
Abcam TripleStain IHC Kit: R&R&M on human tissue (DAB, AP/Red & Green/HRP), 18ML
Supplier:  Abcam AB18328818ML
Encompass_Preferred
Abcam's TripleStain IHC Kit (ab183288) is designed to be used with one mouse and two rabbit primary antibodies, supplied by the user, to detect three distinct antigens on human tissue or cell samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-259-0561