Learn More
TARGETMOL CHEMICALS INC TRIPLIN 5MG
Supplier: TARGETMOL CHEMICALS INC T349565MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Triplin is a chelator of copper ion. Triplin affects the transport of copper ions from ATX1 to RAN1. purity: 98%