Learn More
AdipoGen Tripolin B (1 mg)
SDP
Supplier:  AdipoGen AGCR13521M001
Encompass_Preferred
Chemical. CAS: 372164-71-1. Formula: C12H9N3O. MW: 211.2. Aurora A and B modulator.
Catalog No. 50-205-3905