Learn More
Selleck Chemical LLC Triprolidine Hydrochloride S5447-25mg
SDP
Supplier:  Selleck Chemical LLC S544725MG
Encompass_Preferred
Triprolidine Hydrochloride is the hydrochloride salt form of Triprolidine which is the first generation histamine H1 antagonist used in allergic rhinitis
Catalog No. 50-311-0337