Learn More
TARGETMOL CHEMICALS INC TRIPTOLIDE 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T217910MG
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Triptolide (PG490) is a diterpenes trioxide immunosuppressive agent extracted from the Chinese herb Tripterygium wilfordii. purity: 99%
Catalog No. 50-258-1003