Learn More
Apexbio Technology LLC Triptolide, 38748-32-2, MFCD00210565, 10 mg
SDP
Supplier:  Apexbio Technology LLC A389110
Encompass_Preferred
MF: C20H24O6, MW: 360.41, Synonyms: PG490 ;PG 490 ;PG-490. Solubility: 36 mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. Triptolide is the major bioactive constituent extracted from the Chinese herb Tripterygium wilfordii. Triptolide inhibit the expression of IL-2 in activated T cells and NF-B mediated transcription activation. Triptolide also can inhibit colony formation and the proliferation of tumor cells at extremely low concentrations.
Catalog No. 50-101-1027